Anti-HER2 Tyr1222 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-13091R-A750
BOSSBS-13091R-A750EA 860 GBP
BOSSBS-13091R-A750
Anti-HER2 Tyr1222 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 ofMore Product Information