Anti-SLC1A6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-13046R-A647
BOSSBS-13046R-A647EA 838 GBP
BOSSBS-13046R-A647
Anti-SLC1A6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Excitatory Amino Acid Transporters (EAATs) are membrane-bound proteins that are localized in glial cells and pre-synaptic glutamatergic nerve endings. EAATs transport the excitatory neurotransmitters L-glutamate and D-aspartate, a process that is essential for terminating the postsynaptic action of glutamate. The re-uptake of amino acid neurotransmitters by EAAT proteins has been shown to protect neurons from excitotoxicity, which is caused by the accumulation of amino acid neurotransmitters. EAAT4 is an aspartate/glutamate transporter that is expressed predominantly in the cerebellum. TheMore Product Information