Anti-CTBS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-12937R-HRP
BOSSBS-12937R-HRPEA 844 GBP
BOSSBS-12937R-HRP
Anti-CTBS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
CTBS is an evolutionarily conserved member of the glycosyl hydrolase 18 family of proteins. Localizing to the lysosome, CTBS plays a role in the degradation of asparagine-linked (Asn-linked) glycoproteins. Glycoproteins are translocated to lysosomes via endocytosis or autophagy where they are broken down by proteases and glycosidases. The catabolism of glycoproteins is an important step in the regular turnover of cellular contents and in maintaining the homeostasis of glycosylation. CTBS functions as a glycosidase that cleaves the reducing end GlcNAc from the core chitobiase unit ofMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CTBS
Antigen name chitobiase, di-N-acetyl-
Environmentally Preferable
Antigen synonyms CTB
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1486
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR