Anti-GDF8 Goat Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-1288G-A350
BOSSBS-1288G-A350EA 1270 GBP
BOSSBS-1288G-A350
Anti-GDF8 Goat Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Myostatin (GDF8)is expressed uniquely in human skeletal muscle as a 12 kDa mature glycoprotein consisting of 113 amino acid residues and secreted into plasma. Myostatin is a member of the transforming growth factor beta superfamily of secreted growth and differentiation factors that is essential for proper regulation of skeletal muscle mass. Studies have shown that myostatin could play an important role in cardiac development and physiology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GDF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Goat
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR