Anti-ATP6V1B2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-12549R-A555
BOSSBS-12549R-A555EA 838 GBP
BOSSBS-12549R-A555
Anti-ATP6V1B2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A–H) make up the V1 domain and five subunits (a, d, c, c' and c") make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol ATP6V1B2
Environmentally Preferable
Antigen name ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2
Antigen synonyms ATP6B1B2|HO57|VATB|Vma2|ATP6B2|VPP3
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 526
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR