Anti-ZNF342 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-12212R-A647
BOSSBS-12212R-A647EA 838 GBP
BOSSBS-12212R-A647
Anti-ZNF342 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Krüppel C2H2-type zinc-finger protein family, ZNF342 (zinc finger protein 342), also known as Zinc finger protein 296, is a 475 amino acid nuclear protein that contains six C2H2-type zinc fingers through which it is thought toMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ZNF342
Antigen name Zinc Finger Protein 342
Environmentally Preferable
Antigen synonyms ZN296_HUMAN.|ZNF296|Zinc finger protein 296|zinc finger protein 342
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 162979
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR