Anti-STRN3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-12135R-A488
BOSSBS-12135R-A488EA 838 GBP
BOSSBS-12135R-A488
Anti-STRN3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Striatin, SG2NA, and zinedin, the three mammalian members of the striatin family, are multimodular, WD repeat and calmodulin-binding proteins. Zinedin and SG2NA share with striatin identical protein-protein interaction domains and the same overall domain structure. All three proteins are both cytosolic and membrane-bound and bind calmodulin in the presence of calcium. Striatin is a neuronal, intracellular protein strictly expressed in the somato-dendritic compartment, including spines and subsets of neurons, and is considered as a marker of neuronal polarity. Downregulation of striatin,More Product Information