Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11892R-CY7
BOSSBS-11892R-CY7EA 844 GBP
BOSSBS-11892R-CY7
Anti-NAV2/RAINB1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Neuron navigator 2 (NAV2), also known as RAINB1 (retinoic acid inducible in neuroblastoma 1), POMFIL2 (pore membrane and/or filament-interacting-like protein 2), HELAD1 (helicase APC down-regulated 1), unc53H2 or steerin-2, is a 2,488 amino acid nuclear protein that plays a role in the development of sensory organs. Existing as thirteen alternatively spliced isoforms, Neuron navigator 2 displays 3' to 5' helicase activity and exonuclease activity and participates in the catalysis of ATP to ADP. Neuron navigator 2 is highly expressed in the nervous system of developing embryos and colonMore Product Information