Anti-SCGN Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11754R-CY5
BOSSBS-11754R-CY5EA 844 GBP
BOSSBS-11754R-CY5
Anti-SCGN Rabbit Polyclonal Antibody (Cy5®)
Antibodies
SCGN is a 276 amino acid cytoplasmic protein that contains six EF-hand domains and is related to the calicium-binding proteins Calretinin and Calbindin D28K. Expressed in a variety of tissues including stomach, thyroid, colon, brain and neuroendocrine cells, SCGN is thought to be involved in cell proliferation and KCl (potassium chloride)-mediated calcium flux events. Through its interaction with KCl and its subsequent ability to modulate calcium storage pools within the cell, SCGN may function to negatively control growth and differentiation rates and, thus, indirectly inhibit cellMore Product Information