Anti-SSA reductase/AKR7A2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11706R-A750
BOSSBS-11706R-A750EA 835 GBP
BOSSBS-11706R-A750
Anti-SSA reductase/AKR7A2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
The aldo-keto reductase 7 (AKR7) family includes AKR7A2, AKR7A3 and AKR7A4 in human, AKR7A5 in mouse and AKR7A2 in rat, all of which function in the metabolism of aflatoxin B(1) and other dicarbonyl-containing compounds. More specifically, AKR7A proteins are involved in the metabolism of compounds with ketone groups on adjacent carbon atoms in a broad range of tissues, notably the liver. The human AKR7A2 gene maps to human chromosome 1p35-36, a region frequently deleted in sporadic colorectal cancer. The functional significance of this correlation lies in the constitutive expression of AKR7A2More Product Information