Anti-GALR3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11528R-A750
BOSSBS-11528R-A750EA 835 GBP
BOSSBS-11528R-A750
Anti-GALR3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
GALR3 a 368 and 370 amino acid protein in human and rat, respectively, belongs to a family of G protein-coupled receptors that bind the neuropeptide galanin, which is distributed throughout the central and peripheral nervous system, the pituitary gland, the gastrointestinal tract and in the endocrine and exocrine pancreas. GALR3 mRNA is widely distributed, but expressed at low abundance. In human, GALR3 mRNA is highly expressed in the hypothalamus, pituitary and testis, and is expressed to a lesser extent in adrenal gland and pancreas. Rat and human GALR3 co-express with potassium channelMore Product Information