Anti-Synaptotagmin 1+2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-1136R-A680
BOSSBS-1136R-A680EA 835 GBP
BOSSBS-1136R-A680
Anti-Synaptotagmin 1+2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).[supplied by OMIM].

Type: Primary
Antigen: synaptotagmin 1+2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 285-380/419
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR