Anti-ICA1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11349R-A680
BOSSBS-11349R-A680EA 835 GBP
BOSSBS-11349R-A680
Anti-ICA1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Carbonic anhydrases (CAs), also designated carbonate dehydratases or carbonate hydrolyases, form a large family of genes that encode zinc metalloenzymes of great physiologic importance. As catalysts of the reversible hydration of carbon dioxide, these enzymes participate in a variety of biologic processes, including respiration, acid-base balance, bone resorption and calcification as well as the formation of aqueous humor, cerebrospinal fluid, saliva and gastric acid. Genes in the carbonic anhydrase family encode either active carbonic anhydrase isozymes or _catalytic?(devoid of CO2More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name ICA1
Antigen synonyms OTTHUMP00000200933|OTTHUMP00000200993|Islet cell autoantigen 1 isoform|Islet cell autoantigen 1|ICA69_HUMAN|Islet cell autoantigen 1 69kD|ICA69|Islet cell autoantigen 1 69kDa|OTTHUMP00000200941|p69|OTTHUMP00000200934|ICA 1|69 kDa islet cell autoantigen|Diabetes mellitus type I autoantigen|ICAp69|Islet cell autoantigen p69|ICA1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 1-100/483
Immunogen KLH conjugated synthetic peptide derived from human ICA1
Form Liquid
Molecular weight 55 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR