Anti-JunB Thr102 + Thr104 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11228R-A750
BOSSBS-11228R-A750EA 860 GBP
BOSSBS-11228R-A750
Anti-JunB Thr102 + Thr104 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Transcription factor involved in regulating gene activity following the primary growth factor response. Binds to the DNA sequence 5'-TGA[CG]TCA-3'.

Type: Primary
Antigen: JunB Thr102 + Thr104
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR