Anti-CRP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-0155R-A555
BOSSBS-0155R-A555EA 845 GBP
BOSSBS-0155R-A555
Anti-CRP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Displays several functions associated with host defense: it promotes agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. Can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR