Safety spectacles, Tryon
Supplier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TRYOFLASH TRYOPSF TRYOPSI TRYOCSP
BOLETRYOFLASHEA 0 GBP
BOLETRYOFLASH BOLETRYOPSF BOLETRYOPSI BOLETRYOCSP
Safety spectacles, Tryon
Safety Glasses
Ultra-wrap-around, sporty design safety spectacles with co-injected FLEX 160° temples, adjustable and non-slip bridge and plantium coating.
  • Wrap-around frame
  • Platinum coating - except flash version
  • Co-injected PC/TPR temples
  • Non-slip adjustable bridge
  • Flex 160° temples
The permanent coating applied on both surfaces makes them highly scratch-resistant gives them high resistance to the most aggressive chemicals and slows the appearance of fogging. In any circumstances and at every moment, Platinum® provides the highest safety for eyes.
Order Now
SPECIFICATIONS
Environmentally Preferable