Safety Glasses, NESS+ Small
Supplier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NESSPCSP NESSPPSI NESSPPSF PSSNESF028 NESPSN10E NESPSN30E
BOLENESSPCSPEA 10.12 GBP
BOLENESSPCSP BOLENESSPPSI BOLENESSPPSF BOLEPSSNESF028 BOLENESPSN10E BOLENESPSN30E
Safety Glasses, NESS+ Small
Safety Glasses
With Ness+ you enjoy all the benefits of the NESS model with added comfort. Exclusive PLATINUM scratch and fog resistant coating helps you work more efficiently. Ness+ is also available in two sizes.
  • Ultra wrap-around frame for perfect protection
  • Non-slip bridge
  • Bi-material temples for better comfort
  • PLATINUM coating: anti-scratch and anti-fog