Desiccator cabinets, auto, SICCO Maxi series
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
V1914-01 V1916-01
BOHLV1914-01EA 9410 GBP
BOHLV1914-01 BOHLV1916-01
Desiccator cabinets, auto, SICCO Maxi series
Desiccators Desiccator Cabinets
Desiccator cabinets, transparent acrylic, aluminium frame with panels made of acrylic glass, which are suitable for long-term storage.
  • Door with magnetic catch and circular rubber seal
  • Four casters (two of the casters with brakes)
  • Variable height telescopic shelves made of stainless steel
These desiccator can maintain a constant humidity of between 20 and 30%. They are the ideal solution for storing reference samples, electronics, photographic equipment, papers and valuable cultural assets.