Surface protection sheets, BOLA Sheets PTFE or FEP
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S1803-02 S1803-04 S1803-06 S1803-08 S1803-14 S1803-23 S1803-25 S1833-04 S1803-12 S1803-31 S1803-29 S1803-27 S1803-21 S1803-10 S1833-46 S1833-38 S1833-34 S1833-16 S1833-08 S1803-33
BOHLS1803-02EA 17.1 GBP
BOHLS1803-02 BOHLS1803-04 293-6112 293-6111 BOHLS1803-14 BOHLS1803-23 BOHLS1803-25 BOHLS1833-04 BOHLS1803-12 BOHLS1803-31 BOHLS1803-29 BOHLS1803-27 BOHLS1803-21 BOHLS1803-10 BOHLS1833-46 BOHLS1833-38 BOHLS1833-34 BOHLS1833-16 BOHLS1833-08 BOHLS1803-33
Surface protection sheets, BOLA Sheets PTFE or FEP
Surface Protectors Bench Protectors
Protection sheets made from PTFE or FEP.
  • PTFE rolls in white
  • FEP rolls in transparent, gastight and non-porous
  • Temperature resistant from −200 to +250 °C
For use as table pad or lining drawers. Also suitable for using as slideway or for insulation.
Delivery information: Delivered on rolls.