PFA-EX tubings, flexible, BOLA
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S1824-60 S1824-63 S1824-57 S1824-54 S1824-33 S1824-30 S1824-27 S1824-24 S1824-80 S1824-74 S1824-77 S1824-83
BOHLS1824-60EA 362 GBP
BOHLS1824-60 BOHLS1824-63 BOHLS1824-57 BOHLS1824-54 BOHLS1824-33 BOHLS1824-30 BOHLS1824-27 BOHLS1824-24 BOHLS1824-80 BOHLS1824-74 BOHLS1824-77 BOHLS1824-83
PFA-EX tubings, flexible, BOLA
Tubing Transfer Tubing
Conductive corrugated tubing with nominal width and circular corrugations around the longitudinal axis.
  • Can be connected directly to fittings, stopcocks or hose connectors
  • High temperature resistance
Applications include ideal for connections under vibrations, easy handling of liquids and gases, transport of solvents or alcohols, for compensation of thermal expansions and antistatic applications. It has good bending radius and burst pressure.
Order Now
SPECIFICATIONS
Environmentally Preferable