Waste Tubing Desktop Mounts, b.safe Multiport II
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R1125-02
BOHLR1125-02EA 315 GBP
BOHLR1125-02
Waste Tubing Desktop Mounts, b.safe Multiport II
Tubing Transfer Tubing
One connection for collecting liquid waste from up to 2 HPLC systems that can be integrated directly in laboratory hoods or working tables. b.safe Waste Tubes lead liquid solvent waste directly into connected canisters.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways
  • Additional slope improves the flow and prevents any backflow
Made of static dissipative PE-EX, suitable for boards with a thickness of 10 to 35 mm, easy fixing by counter nut. Low installation height due toMore Product Information