Filtering tiles, BOLA
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N1610-10 N1616-30 N1616-20 N1616-10
BOHLN1610-10EA 827 GBP
BOHLN1610-10 BOHLN1616-30 BOHLN1616-20 BOHLN1616-10
Filtering tiles, BOLA
Filters Filter Tiles
These filtering tiles are made of microporous PTFE.
  • Suitable for cutting or stamping
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C