Tweezers, BOLA
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H909-04 H912-02 H912-04 H912-06 H909-06 H909-02
BOHLH909-04EA 35.8 GBP
BOHLH909-04 BOHLH912-02 BOHLH912-04 BOHLH912-06 BOHLH909-06 BOHLH909-02
Tweezers, BOLA
Forceps
Precast tweezers made of PTFE.
  • With pointed or blunt ends
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C