Ground joint tube fittings, for pressure upto 5 Bar, BOLA
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H1001-04 H1001-14 H1001-12 H1001-10 H1001-06
BOHLH1001-04EA 55.9 GBP
BOHLH1001-04 BOHLH1001-14 BOHLH1001-12 BOHLH1001-10 BOHLH1001-06
Ground joint tube fittings, for pressure upto 5 Bar, BOLA
Adapters Adapters, Joints with Tube Connectors
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
  • Vacuum: suitable
Fitting made of PTFE for transition from ground joints to metric threads for connecting hard-walled tubing (e.g. PTFE, PFA or FEP). With nuts made of glass-fibre reinforced PTFE, body with rings and knurled grip for opening. The product is only exposed to PTFE.
Order Now
SPECIFICATIONS
Environmentally Preferable