Hose connectors, with nut
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D581-02 D581-04 D582-04 D581-06 D582-02 D581-08
BOHLD581-02EA 10 GBP
BOHLD581-02 BOHLD581-04 BOHLD582-04 BOHLD581-06 BOHLD582-02 BOHLD581-08
Hose connectors, with nut
Fittings and Connectors
GL 14 and GL 18 hose connectors, made of PFA, GL 25 and GL 32 hose connectors, made of PTFE, with elastic sealing lip and FPM O-ring and nut made of PPS.
  • Available as straight or bent type
  • Universal chemical resistance
  • Maximum temperature up to 250 °C