Stirrer blades
Supplier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C440-08 C446-12 C440-10 C443-10 C441-10 C443-14 C448-10 C446-10 C443-08 C446-08 C448-08 C441-12 C448-42 C448-36 C448-28 C448-20 C446-16 C446-14 C441-14 C441-16 C442-08 C442-10
BOHLC440-08EA 147 GBP
BOHLC440-08 BOHLC446-12 BOHLC440-10 BOHLC443-10 BOHLC441-10 BOHLC443-14 BOHLC448-10 BOHLC446-10 BOHLC443-08 BOHLC446-08 BOHLC448-08 BOHLC441-12 BOHLC448-42 BOHLC448-36 BOHLC448-28 BOHLC448-20 BOHLC446-16 BOHLC446-14 BOHLC441-14 BOHLC441-16 BOHLC442-08 BOHLC442-10
Stirrer blades
Stirrers Stirring Blades
Stirrer blades made from PTFE, with a set of clamp screws made from a PTFE/PEEK compound.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Create axial flow
The blades can be fixed tightly on stirrer shafts by means of the clamp screws. A spanner wrench is included for easy assembly.