Urine collection container with or without snap seals
Supplier: BOETTGER PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06.023.0000 06.013.0100
BOET06.023.0000EA 271 GBP
BOET06.023.0000 BOET06.013.0100
Urine collection container with or without snap seals
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP or LDPE, translucent.
  • Resistant to pressure and shatterproof
  • Optimal visibility of content, graduated
  • Fluid tight lid, easy to close
  • Frosted surface on cups and lids for identification marking
Ideal for collecting and storage of urine samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904