Microplate shakers, Jitterbug
Supplier: BOEKEL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
270400 270440
BOEI270400EA 5170 GBP
BOEI270400 BOEI270440
Microplate shakers, Jitterbug
Shakers Microplate Shakers
Designed to suitable for two or four standard microplates or strip trays.
  • Calibration 'Mode'
  • Temperature calibration
  • Heated lid to provide greater uniformity
  • Independently set temperature, shaking speed and mixing duration