Dressing jars, with lid
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8760 8761 8771 8770 8772 8762
BOCH8760EA 27.9 GBP
BOCH8760 BOCH8761 213-0246 BOCH8770 213-0247 BOCH8762
Dressing jars, with lid
Jars General Use Jars
Dressing jar either with flat or high shape, made of aluminium.
  • Very good temperature resistance
  • Good chemical resistance
  • Lightweight
Order Now
SPECIFICATIONS
Environmentally Preferable