Bowls
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8671 8677 8670 8672 8673 8674
BOCH8671EA 55.1 GBP
BOCH8671 BOCH8677 BOCH8670 BOCH8672 BOCH8673 BOCH8674
Bowls
Trays/Dishes/Pots Laboratory Trays/Dishes
Made of stainless steel with flat bottom.
  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean