Instrument trays
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8484 8486 8480 8485
BOCH8484EA 138 GBP
BOCH8484 BOCH8486 BOCH8480 BOCH8485
Instrument trays
Trays/Dishes/Pots Instrument Trays/Dishes
Made of 18/10 steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sturdy