Crucibles, metal
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8102 8106 8127 8881 8882 8883 8122 8124 8126 8100 8123 8129 8105 8103 8101 8109 8125 8145 8107 8104
BOCH8102EA 96.5 GBP
BOCH8102 BOCH8106 459-1194 BOCH8881 BOCH8882 459-0154 459-1190 459-1192 459-1193 BOCH8100 BOCH8123 BOCH8129 BOCH8105 BOCH8103 BOCH8101 BOCH8109 BOCH8125 BOCH8145 BOCH8107 BOCH8104
Crucibles, metal
Crucibles
These crucibles and lids are made of nickel or stainless steel 18/10.
Ordering information: Lids need to be ordered separately.