Retort rods, with M10 thread
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5110 5111 5112 5113 5114 5170 5171 5116 5172 5173 5175 5174 5115 5117 5118
BOCH5110EA 39.8 GBP
BOCH5110 BOCH5111 BOCH5112 BOCH5113 BOCH5114 BOCH5170 BOCH5171 BOCH5116 BOCH5172 BOCH5173 BOCH5175 BOCH5174 BOCH5115 BOCH5117 BOCH5118
Retort rods, with M10 thread
Retort Rods
Stainless steel 18/10.
  • Rods for stand bases
  • M10 thread