Stand bases
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5031 5030 5032 5033 5034 5037 5038 5035 5036
BOCH5031EA 60.2 GBP
BOCH5031 BOCH5030 BOCH5032 241-3167 BOCH5034 BOCH5037 BOCH5038 BOCH5035 BOCH5036
Stand bases
Retort Stands
Steel, varnished.
  • With M10 threaded hole
  • One adjustable foot
Certifications: DIN 12892