Dishes, disposable
Supplier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11615 11610 11611 11613
BOCH11615EA 131 GBP
BOCH11615 BOCH11610 BOCH11611 BOCH11613
Dishes, disposable
Boats Weighing Boats
Aluminium.
  • Flat bottom and crimped sides for rigidity
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes feature crimped sides. Ideal for general-purpose weighing, dispensing, and storage applications. Dishes also function as evaporating dishes, parts containers, or dust covers.