Anti-KRT76 Mouse Monoclonal Antibody (Purified) [clone: KRTH/1076]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BNUB1076-500 BNUB1076-100
BNUB1076-500EA 487 GBP
BNUB1076-500 BNUB1076-100
Anti-KRT76 Mouse Monoclonal Antibody (Purified) [clone: KRTH/1076]
Antibodies
This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: KRTH/1076
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR