Anti-INS Mouse Monoclonal Antibody (Purified) [clone: IRDN/794]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BNUB0794-500 BNUB0794-100
BNUB0794-500EA 487 GBP
BNUB0794-500 BNUB0794-100
Anti-INS Mouse Monoclonal Antibody (Purified) [clone: IRDN/794]
Antibodies
Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is theMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol INS
Antigen name Insulin
Environmentally Preferable
Antigen synonyms MODY10|ILPR|IDDM2|Proinsulin|IRDN
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone IRDN/794
Reactivity Pig,Human,Bovine,Rat,Mouse
Host Mouse
Gene ID 272259
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR