Anti-INS Mouse Monoclonal Antibody (Purified) [clone: 2D11-H5]
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNUB0030-500 BNUB0030-100
BNUB0030-500EA 487 GBP
BNUB0030-500 BNUB0030-100
Anti-INS Mouse Monoclonal Antibody (Purified) [clone: 2D11-H5]
Antibodies
Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is theMore Product Information