Anti-VIM Mouse Monoclonal Antibody (CF640R) [clone: VM1170]
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNC401170-500 BNC401170-100
BNC401170-500EA 533 GBP
BNC401170-500 BNC401170-100
Anti-VIM Mouse Monoclonal Antibody (CF640R) [clone: VM1170]
Antibodies
This MAb reacts with a 58 kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFP s) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and SchwannomasMore Product Information