Imidazole

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

54556-1 54556-2 54556-3
BMOL54556-1EA 0 GBP
BMOL54556-1 BMOL54556-2 BMOL54556-3
Imidazole
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR