Nonyl-β-D-glucopyranoside

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol, N-Nonyl Beta-D-glucopyranoside, β-Nonylglucoside, Nonyl β-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(nonyloxy)tetrahydro-2H-pyran-3,4,5-triol, NG

50862-1 50862-2
BMOL50862-1EA 0 GBP
BMOL50862-1 BMOL50862-2
Nonyl-β-D-glucopyranoside
Nonyl β-D-glucopyranoside
Formula: C₁₅H₃₀O₆
MDL Number: MFCD00063300
CAS Number: 69984-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR