Octyl-β-D-thioglucopyranoside

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

50739-2 50739-3
BMOL50739-2EA 0 GBP
BMOL50739-2 BMOL50739-3
Octyl-β-D-thioglucopyranoside
Octyl β-D-thioglucopyranoside
Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR