Bis-trispropane

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]

50224-1 50224-2
BMOL50224-1EA 0 GBP
BMOL50224-1 BMOL50224-2
Bis-trispropane
BIS-TRIS propane
Formula: C₁₁H₂₆N₂O₆
MDL Number: MFCD00004689
CAS Number: 64431-96-5
EINECS: 264-899-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR