Acrylamide, crystallized_4

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Prop-2-enamide

50161-1 50161-2
BMOL50161-1EA 0 GBP
BMOL50161-1 BMOL50161-2
Acrylamide, crystallized_4
Acrylamide (monomer)
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 232
Melting Pt: 84.5
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR