D-(+)-Maltose monohydrate

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

05957-1 05957-2
BMOL05957-1EA 0 GBP
BMOL05957-1 BMOL05957-2
D-(+)-Maltose monohydrate
D(+)-Maltose monohydrate
Formula: C₁₂H₂₂O₁₁·1H₂O
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR