Glycylglycine (Gly-Gly)

Supplier: BIOMOL RESEARCH LABORATORIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

05008-1 05008-2
BMOL05008-1EA 0 GBP
BMOL05008-1 BMOL05008-2
Glycylglycine (Gly-Gly)
Proteins and Peptides

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Order Now

SPECIFICATIONS

Protein/peptide name Glycylglycine (Gly-Gly)
Protein synonyms Glycylglycine-Gly-Gly
Environmentally Preferable
Species
Source
CAS No. 556-50-3
MDL No. MFCD00008130
Molecular weight 132,12 g/mol
Molecular formula C₄H₈N₂O₃
Storage conditions Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR