Human bone marrow, unprocessed
Supplier: Charles River Laboratories Cell Solutions
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BM008F-3 BM008F-4
BM008F-3EA 0 GBP
BM008F-3 BM008F-4
Human bone marrow, unprocessed
Cells
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
Caution: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.