Reagent reservoirs, disposable
Supplier: BIOTIX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SR-0050-1SC SR-0025-BWM 63300130 63300133 63300135 63300120 63300121 63300126 63300129 63300124 63300127 63300125 63300128 63300118
613-1189EA 0 GBP
613-1189 613-2857 613-1190 613-1193 613-1191 613-2852 613-2853 BITX63300126 613-2860 BITX63300124 BITX63300127 613-2856 613-2859 613-2850
Reagent reservoirs, disposable
Reagent Reservoirs
These disposable reagent reservoirs are designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes. Users can choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
  • Sterile or non sterile
  • Angled sides and a V-shaped bottom allow maximum recovery of reagents
  • All reservoirs feature internal graduated volume lines and pour-spouts on the corners
  • Certified free of DNA, DNase, RNase and endotoxins (pyrogens)
Manufactured under stringent controls inMore Product Information