Anti-LZTS2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB89898-50
BIRBORB89898-50EA 0 GBP
BIRBORB89898-50
Anti-LZTS2 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: LZTS2 (leucine zipper, putative tumor suppressor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR