Anti-TET3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ORB67241-100
BIRBORB67241-100EA 0 GBP
BIRBORB67241-100
Anti-TET3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunoprecipitation: 1:50-1:100

Type: Primary
Antigen: TET3 (tet methylcytosine dioxygenase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR